Assay Details
| Gene Symbol: CCDC189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195631.1 | NP_001182560.1 | 8 - 9 | 895 | 59 | |
| GenBank mRNA | BC149051.1 | - | 6 - 7 | 1204 | 59 | |
More Information
Set Membership: |
|

CCDC189
Genomic Map