Assay Details
| Gene Symbol: GDF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001140.1 | NP_001001140.1 | 1 - 2 | 418 | 102 | |
| GenBank mRNA | U13661.1 | - | 1 - 2 | 325 | 102 | |
More Information
Set Membership: |
|

GDF6
Genomic Map