Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010996.1 | NP_001010996.1 | 2 - 3 | 214 | 121 | |
| GenBank mRNA | AY860499.1 | - | 2 - 3 | 214 | 121 | |
| BC123902.1 | - | 2 - 3 | 243 | 121 | ||
More Information
Set Membership: |
|

FCN1
Genomic Map