Assay Details
| Gene Symbol: BOLA-DYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012679.1 | NP_001012697.1 | 5 - 6 | 839 | 87 | |
| GenBank mRNA | AJ580584.1 | - | 5 - 6 | 839 | 87 | |
More Information
Set Membership: |
|

BOLA-DYB
Genomic Map