Assay Details
| Gene Symbol: BOLA-DOB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013600.1 | NP_001013618.1 | 4 - 5 | 897 | 100 | |
| GenBank mRNA | AB117945.1 | - | 4 - 5 | 758 | 100 | |
| AB117946.1 | - | 4 - 5 | 897 | 100 | ||
More Information
Set Membership: |
|

BOLA-DOB
Genomic Map