Assay Details
| Gene Symbol: B3GAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014392.1 | NP_001014414.1 | 3 - 4 | 892 | 71 | |
| GenBank mRNA | AJ890840.1 | - | 3 - 4 | 892 | 71 | |
More Information
Set Membership: |
|

B3GAT2
Genomic Map