Assay Details
| Gene Symbol: BCL2L14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015544.1 | NP_001015544.1 | 3 - 4 | 791 | 70 | |
| GenBank mRNA | BT020906.1 | - | 3 - 4 | 791 | 70 | |
More Information
Set Membership: |
|

BCL2L14
Genomic Map