Assay Details
| Gene Symbol: PSMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015639.1 | NP_001015639.1 | 4 - 5 | 321 | 68 | |
| GenBank mRNA | BC102391.1 | - | 4 - 5 | 358 | 68 | |
| BT020961.1 | - | 4 - 5 | 321 | 68 | ||
More Information
Set Membership: |
|

PSMC2
Genomic Map