Assay Details
| Gene Symbol: MRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034552.1 | NP_001029724.1 | 3 - 4 | 245 | 97 | |
| GenBank mRNA | BC146146.1 | - | 3 - 4 | 97 | ||
| BT021655.1 | - | 3 - 4 | 245 | 97 | ||
More Information
Set Membership: |
|

MRO
Genomic Map