Assay Details
| Gene Symbol: ASMTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035058.1 | NP_001030230.1 | 9 - 10 | 1257 | 53 | |
| GenBank mRNA | BC102999.1 | - | 9 - 10 | 1257 | 53 | |
More Information
Set Membership: |
|

ASMTL
Genomic Map