Assay Details
| Gene Symbol: ASMTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035058.1 | NP_001030230.1 | 10 - 11 | 1392 | 87 | |
| GenBank mRNA | BC102999.1 | - | 10 - 11 | 1392 | 87 | |
More Information
Set Membership: |
|

ASMTL
Genomic Map