Assay Details
| Gene Symbol: C3H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038026.2 | NP_001033115.1 | 1 - 2 | 81 | 89 | |
| GenBank mRNA | BC109726.1 | - | 1 - 2 | 83 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109726.1, NM_001038026.2 transcripts.
Set Membership: |
|

C3H1orf189
Genomic Map