Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040517.1 | NP_001035607.1 | 1 - 2 | 158 | 53 | |
| GenBank mRNA | BC102731.1 | - | 1 - 2 | 154 | 53 | |
| DV805559.1 | - | 1 - 2 | 159 | 53 | ||
| DV806540.1 | - | 1 - 2 | 174 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001040517.1 transcripts.
Set Membership: |
|

TNNI3
Genomic Map