Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040517.1 | NP_001035607.1 | 6 - 7 | 522 | 74 | |
| GenBank mRNA | BC102731.1 | - | 6 - 7 | 518 | 74 | |
| DV805559.1 | - | 6 - 7 | 523 | 74 | ||
| DV806540.1 | - | 6 - 7 | 511 | 74 | ||
More Information
Set Membership: |
|

TNNI3
Genomic Map