Assay Details
| Gene Symbol: SOCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046182.1 | NP_001039647.1 | 1 - 2 | 219 | 89 | |
| GenBank mRNA | BC114096.1 | - | 1 - 2 | 219 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114096.1, NM_001046182.1 transcripts.
Set Membership: |
|

SOCS5
Genomic Map