Assay Details
| Gene Symbol: MT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075140.1 | NP_001068608.1 | 1 - 2 | 113 | 87 | |
| GenBank mRNA | BC112477.1 | - | 114 | 87 | ||
| DT859958.1 | - | 1 - 2 | 109 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112477.1, NM_001075140.1 transcripts.
Set Membership: |
|

MT2A
Genomic Map