Assay Details
| Gene Symbol: RAB1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076233.1 | NP_001069701.1 | 4 - 5 | 322 | 66 | |
| GenBank mRNA | BC105393.1 | - | 4 - 5 | 322 | 66 | |
More Information
Set Membership: |
|

RAB1B
Genomic Map