Assay Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076413.2 | NP_001069881.1 | 1 - 2 | 201 | 82 | |
| GenBank mRNA | BC118345.1 | - | 1 - 2 | 202 | 82 | |
More Information
Set Membership: |
|

TRAPPC1
Genomic Map