Assay Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197136.1 | NP_001184065.1 | 1 - 2 | 130 | 87 | |
| GenBank mRNA | DN279961.1 | - | 4 - 5 | 569 | 87 | |
| DT849117.1 | - | 1 - 2 | 131 | 87 | ||
More Information
Set Membership: |
|

ZSWIM7
Genomic Map