Assay Details
| Gene Symbol: ZSWIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197136.1 | NP_001184065.1 | 2 - 3 | 148 | 63 | |
| GenBank mRNA | DN279961.1 | - | 3 - 4 | 551 | 63 | |
| DT849117.1 | - | 2 - 3 | 149 | 63 | ||
More Information
Set Membership: |
|

ZSWIM7
Genomic Map