Assay Details
| Gene Symbol: RBM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198892.1 | NP_001185821.1 | 2 - 3 | 208 | 83 | |
| GenBank mRNA | DQ341265.1 | - | 2 - 3 | 208 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198892.1 transcripts.
Set Membership: |
|

RBM12
Genomic Map