Assay Details
| Gene Symbol: EIF4EBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204168.1 | NP_001191097.1 | 2 - 3 | 353 | 80 | |
| GenBank mRNA | EH179060.1 | - | 2 - 3 | 353 | 80 | |
More Information
Set Membership: |
|

EIF4EBP3
Genomic Map