Assay Details
Gene Symbol: P2RY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204449.1 | NP_001191378.1 | 1 - 2 | 107 | 59 | |
GenBank mRNA | DV873354.1 | - | 1 - 2 | 107 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
