Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204851.1 | NP_001191780.1 | 2 - 3 | 2124 | 76 | |
| GenBank mRNA | BI775903.1 | - | 2 - 3 | 368 | 76 | |
More Information
Set Membership: |
|

STON1
Genomic Map