Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205379.1 | NP_001192308.1 | 3 - 4 | 248 | 81 | |
| GenBank mRNA | BC103103.1 | - | 3 - 4 | 209 | 81 | |
More Information
Set Membership: |
|

TEDC2
Genomic Map