Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205379.1 | NP_001192308.1 | 5 - 6 | 705 | 78 | |
| GenBank mRNA | DT808282.1 | - | 2 - 3 | 448 | 78 | |
More Information
Set Membership: |
|

TEDC2
Genomic Map