Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205379.1 | NP_001192308.1 | 9 - 10 | 1204 | 86 | |
| GenBank mRNA | BC103103.1 | - | 9 - 10 | 1186 | 86 | |
| CK849517.1 | - | 1 - 2 | 553 | 86 | ||
More Information
Set Membership: |
|

TEDC2
Genomic Map