Assay Details
| Gene Symbol: C21H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205388.1 | NP_001192317.1 | 2 - 3 | 428 | 106 | |
| GenBank mRNA | BC114110.1 | - | 2 - 3 | 429 | 106 | |
| DV844194.1 | - | 2 - 3 | 429 | 106 | ||
| DV844417.1 | - | 2 - 3 | 428 | 106 | ||
More Information
Set Membership: |
|

C21H14orf180
Genomic Map