Assay Details
| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205409.1 | NP_001192338.1 | 3 - 4 | 377 | 105 | |
| GenBank mRNA | BC149329.1 | - | 3 - 4 | 378 | 105 | |
More Information
Set Membership: |
|

YPEL3
Genomic Map