Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242584.1 | NP_001229513.1 | 1 - 2 | 718 | 110 | |
| GenBank mRNA | BC149113.1 | - | 1 - 2 | 718 | 110 | |
More Information
Set Membership: |
|

IGBP1
Genomic Map