Assay Details
| Gene Symbol: NAA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046511.2 | NP_001039976.1 | 2 - 3 | 224 | 109 | |
| GenBank mRNA | BC112807.1 | - | 2 - 3 | 225 | 109 | |
More Information
Set Membership: |
|

NAA10
Genomic Map