Assay Details
| Gene Symbol: KRT86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193068.1 | NP_001179997.1 | 3 - 4 | 646 | 121 | |
| GenBank mRNA | BC126722.1 | - | 3 - 4 | 463 | 121 | |
More Information
Set Membership: |
|

KRT86
Genomic Map