Assay Details
Gene Symbol: DEFB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102362.2 | NP_001095832.1 | 1 - 2 | 192 | 115 | |
GenBank mRNA | EH162784.1 | - | 1 - 2 | 192 | 115 |
Gene Symbol: DEFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282581.1 | NP_001269510.1 | 1 - 2 | 265 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001102362.2, NM_001282581.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
