Assay Details
| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040562.3 | NP_001035652.1 | 1 | 501 | 86 | |
| GenBank mRNA | BC109790.1 | - | 1 | 620 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HMGB4| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040562.3 | NP_001035652.1 | 1 | 501 | 86 | |
| GenBank mRNA | BC109790.1 | - | 1 | 620 | 86 | |
Set Membership: |
|