Assay Details
| Gene Symbol: H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078089.2 | NP_001071557.1 | 619 | 92 | ||
| GenBank mRNA | BC103043.1 | - | 621 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2AJ| Gene Symbol: H2AJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078089.2 | NP_001071557.1 | 619 | 92 | ||
| GenBank mRNA | BC103043.1 | - | 621 | 92 | ||
Set Membership: |
|