Assay Details
| Gene Symbol: H2AW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098097.2 | NP_001091566.1 | 1 | 1324 | 70 | |
| GenBank mRNA | BC134772.1 | - | 1 | 1325 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2AW| Gene Symbol: H2AW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098097.2 | NP_001091566.1 | 1 | 1324 | 70 | |
| GenBank mRNA | BC134772.1 | - | 1 | 1325 | 70 | |
Set Membership: |
|