Assay Details
| Gene Symbol: PCDHB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103248.1 | NP_001096718.1 | 1 | 1217 | 114 | |
| GenBank mRNA | BC149280.1 | - | 1 | 1217 | 114 | |
More Information
Set Membership: |
|

PCDHB11
Genomic Map