Assay Details
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105640.1 | NP_001099110.1 | 4 | 3619 | 119 | |
| GenBank mRNA | BC153268.1 | - | 4 | 3619 | 119 | |
More Information
Set Membership: |
|

PRRG1
Genomic Map