Assay Details
| Gene Symbol: SST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173960.1 | NP_776385.1 | 2 | 524 | 87 | |
| GenBank mRNA | BC103253.1 | - | 2 | 540 | 87 | |
| M31217.1 | - | 2 | 524 | 87 | ||
More Information
Set Membership: |
|

SST
Genomic Map