Assay Details
| Gene Symbol: XIST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DY130193.1 | - | 5 - 6 | 533 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-421 | MIR421 |
bta-mir-421, mir-421 |
-264.2kb | Sense | Chr. X - 76925575 - 76925659 on NCBI Build ARS-UCD1.2 |
| Downstream | cysteine rich hydrophobic domain 1 | CHIC1 | 121.2kb | Anti-sense | Chr. X - 77312352 - 77359558 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

XIST
Genomic Map