Assay Details
| Gene Symbol: LOC503858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003052.2 | 2 - 3 | 240 | 77 | ||
| GenBank mRNA | Z32813.1 | - | 241 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cytochrome P450, family 19, subfamily A, polypeptide 1 |
CYP19A1 |
CYP19, CYP19P1 |
-25.1kb | Sense | Chr. 10 - 59102240 - 59156607 on NCBI Build ARS-UCD1.2 |
| Downstream | transient receptor potential cation channel subfamily M member 7 | TRPM7 | 458.0kb | Sense | Chr. 10 - 59648210 - 59757063 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

LOC503858
Genomic Map