Assay Details
| Gene Symbol: H19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003958.2 | 3 - 4 | 1347 | 63 | ||
| GenBank mRNA | EF010920.1 | - | 1 - 2 | 122 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | insulin like growth factor 2 | IGF2 | -89.9kb | Sense | Chr. 29 - 49395153 - 49422469 on NCBI Build ARS-UCD1.2 | |
| Downstream | mitochondrial ribosomal protein L23 | MRPL23 | 30.0kb | Anti-sense | Chr. 29 - 49542745 - 49549286 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

H19
Genomic Map