Assay Details
| Gene Symbol: RPS27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024613.1 | 5 - 6 | 572 | 88 | ||
| GenBank mRNA | CK982652.1 | - | 5 - 6 | 535 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tropomyosin 3 | TPM3 | -100.2kb | Anti-sense | Chr. 3 - 16318497 - 16346046 on NCBI Build ARS-UCD1.2 | |
| Downstream |
RAB13, member RAS oncogene family |
RAB13 | 2.3kb | Anti-sense | Chr. 3 - 16449594 - 16457409 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

RPS27
Genomic Map