Assay Details
| Gene Symbol: MIR2421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030876.1 | 1 | 41 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 4 C7orf25 homolog | C4H7orf25 | C7orf25 | -305.5kb | Anti-sense | Chr. 4 - 78144385 - 78147228 on NCBI Build ARS-UCD1.2 |
| Downstream | inhibin subunit beta A | INHBA | 827.1kb | Anti-sense | Chr. 4 - 79279914 - 79300035 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2421
Genomic Map