Assay Details
| Gene Symbol: MIR2299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031036.1 | 1 | 43 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | relaxin family peptide receptor 2 | RXFP2 | -360.5kb | Sense | Chr. 12 - 29212314 - 29274747 on NCBI Build ARS-UCD1.2 | |
| Downstream | heat shock protein family H (Hsp110) member 1 | HSPH1 | 160.7kb | Anti-sense | Chr. 12 - 29796074 - 29819631 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2299
Genomic Map