Assay Details
| Gene Symbol: MIR421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031062.1 | 1 | 40 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-374b | MIR374B |
bta-mir-374b, mir-374b |
-74.0 | Sense | Chr. X - 76925407 - 76925501 on NCBI Build ARS-UCD1.2 |
| Downstream | X (inactive)-specific transcript | XIST | 235.9kb | Sense | Chr. X - 77161577 - 77198299 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR421
Genomic Map