Assay Details
| Gene Symbol: MIR410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031064.1 | 1 | 38 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-369 | MIR369 |
MIRN369, bta-mir-369 |
-256.0 | Sense | Chr. 21 - 65954188 - 65954256 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-656 | MIR656 | bta-mir-656 | 743.0 | Sense | Chr. 21 - 65955335 - 65955413 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR410
Genomic Map