Assay Details
| Gene Symbol: MIR429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031066.1 | 1 | 39 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tubulin tyrosine ligase like 10 | TTLL10 | -4.9kb | Sense | Chr. 16 - 51299039 - 51316585 on NCBI Build ARS-UCD1.2 | |
| Downstream | microRNA mir-200a | MIR200A |
MIRN200A, bta-mir-200a, mir-200a |
1.2kb | Sense | Chr. 16 - 51322680 - 51322765 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR429
Genomic Map