Assay Details
| Gene Symbol: MIR2356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031076.1 | 1 | 38 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tensin 1 | TNS1 | TNS | Sense | Chr. 2 - 105902077 - 106118407 on NCBI Build ARS-UCD1.2 | |
| Downstream | RUN and FYVE domain containing 4 | RUFY4 | 142.7kb | Anti-sense | Chr. 2 - 106154128 - 106183766 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2356
Genomic Map