Assay Details
| Gene Symbol: MIR2352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031078.1 | 1 | 38 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | integrin subunit alpha 6 | ITGA6 | Sense | Chr. 2 - 24091589 - 24179355 on NCBI Build ARS-UCD1.2 | ||
| Downstream | distal-less homeobox 2 | DLX2 | 292.3kb | Anti-sense | Chr. 2 - 24448625 - 24450898 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2352
Genomic Map